Learn More
Bellco Glass, Inc. Borosil Tubes, Culture, Round Bottom, Amber, PTFE-Lined PP Screw Caps, 60mL
SDP
Catalog No. 502549869
Encompass_Preferred
Amber Culture Tubes Media Round Bottom PP cap with Liner 60 mL Borosilicate CS/50SpecificationsMaterial: 3.3 Borosilicate Glass Tube size: 60 mLColor: AmberCap Material: Polypropylene Overall Dimension: 9901013Class/Quality Grade: Type I, Class A
Catalog No. 50-254-9869 Supplier Bellco Glass, Inc. Supplier No. 452800893
Amber Culture Tubes Media Round Bottom PP cap with Liner 60 mL Borosilicate CS/50SpecificationsMaterial: 3.3 Borosilicate Glass Tube size: 60 mLColor: AmberCap Material: Polypropylene Overall Dimension: 9901013Class/Quality Grade: Type I, Class A