Learn More
Bellco Glass, Inc. Borosil Tubes, Test, Reusable, Plain End, 55mL, 25mm x 150mm (OD x H),
SDP
Catalog No. 502549554
Encompass_Preferred
Test Tubes without RIM 25*150 Borosilicate 3.3 ISO 4142 55ml CS/200SpecificationsMaterial: GlassVolumen: 55 mLOverall Dimension: 25mm x 150mmBorosilicate Glass 3.3Graduation Range: Type I
Catalog No. 50-254-9554 Supplier Bellco Glass, Inc. Supplier No. 452801208
Test Tubes without RIM 25*150 Borosilicate 3.3 ISO 4142 55ml CS/200SpecificationsMaterial: GlassVolumen: 55 mLOverall Dimension: 25mm x 150mmBorosilicate Glass 3.3Graduation Range: Type I