Learn More
Bellco Glass, Inc. Borosil Tubes, Test, Reusable, Plain End, 15mL, 15mm x 150mm (OD x H)
SDP
Catalog No. 502549557
Encompass_Preferred
Borosil Test Tubes with Rim, Borosilicate 3.3 with Round Bottom, ISO/DIN 4142, 15 mL, CS/400Borosil Test Tubes with Rim comply with IS 2618, ISO/ DIN 4142. Each tube is made of 3.3 Borosilicate glass and is designed with a rim and round bottom. Made thick-walled, mechanically very resistant. The Borosil Test Tubes also have a high thermal shock resistance.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 15 mLApprox O.D. x Length: 15mm x 150mmClass/ Quality Grade: Type IQuantity Per Case: 400
Catalog No. 50-254-9557 Supplier Bellco Glass, Inc. Supplier No. 452801205
Borosil Test Tubes with Rim, Borosilicate 3.3 with Round Bottom, ISO/DIN 4142, 15 mL, CS/400Borosil Test Tubes with Rim comply with IS 2618, ISO/ DIN 4142. Each tube is made of 3.3 Borosilicate glass and is designed with a rim and round bottom. Made thick-walled, mechanically very resistant. The Borosil Test Tubes also have a high thermal shock resistance.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 15 mLApprox O.D. x Length: 15mm x 150mmClass/ Quality Grade: Type IQuantity Per Case: 400