Learn More
Bellco Glass, Inc. Borosil Tubes, Test, Reusable, Plain End, 13mL, 15mm x 125mm (OD x H)
SDP
Catalog No. 502549558
Encompass_Preferred
Borosil Test Tubes without RIM 15*125 Borosilicate 3.3 ISO 4142 13ml CS/800SpecificationsMaterial: GlassVolumen: 13 mLOverall Dimension: 15mm x 125mmBorosilicate Glass 3.3Graduation Range: Type I
Catalog No. 50-254-9558 Supplier Bellco Glass, Inc. Supplier No. 452801204
Borosil Test Tubes without RIM 15*125 Borosilicate 3.3 ISO 4142 13ml CS/800SpecificationsMaterial: GlassVolumen: 13 mLOverall Dimension: 15mm x 125mmBorosilicate Glass 3.3Graduation Range: Type I