Learn More
Bellco Glass, Inc. Borosil Tubes, Test, Reusable, Beaded Rim, 5mL, 12mm x 75mm (OD x H)
SDP
Supplier:  Bellco Glass, Inc. 452801189
Encompass_Preferred
Borosil Test Tubes with Rim, Borosilicate 3.3 with Round Bottom, ISO/DIN 4142, 5 mL, CS/800Borosil Test Tubes with Rim comply with IS 2618, ISO/ DIN 4142. Each tube is made of 3.3 Borosilicate glass and is designed with a rim and round bottom. Made thick-walled, mechanically very resistant. The Borosil Test Tubes also have a high thermal shock resistance.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 5 mLApprox O.D. x Length: 12mm x 75mmClass/ Quality Grade: Type IQuantity Per Case: 800
Catalog No. 50-254-9573