Learn More
Bellco Glass, Inc. Borosil Volumetric Flask With Glass/Plastic Stopper, ASTM E288 Class A, 100 mL
SDP
Supplier:  Bellco Glass, Inc. 452800997
Encompass_Preferred
Borosil Volumetric Flask with Interchangeable Solid Glass/ Plastic Stopper, Accuracy as per Class A, ASTM E288 with Batch Certificate, 100 mL, CS/10Borosil Volumetric Flasks with Interchangeable PP Stoppers are manufactured from 3.3 Borosilicate I.D. control tubing. Each flask is individually calibrated with an automatic, computer controlled, calibration machine, providing a more accurate volume. Borosils Volumetric Flasks are designed to give the best combination of thermal shock resistance, physical strength and resistance to chemical attack. All Volumetric Flasks have permanent amber printing. Comes with Batch Certificate. - 5mL to 20mL supplied with trapezoidal shape for better stability *Certified and Calibrated at 20 C. Material: 3.3 BorosilicateColor: ClearCapacity: 100mLTolerance: 0.08mLNeck Type: Solid Glass/ Plastic StopperNeck Specification: GroundedStopper Size: 14/23Class/ Quality Grade: Type I, Class AQuantity Per Case: 10
Catalog No. 50-254-9765