Learn More
Bellco Glass, Inc. Borosil Watch Glass S-Line Soda Glass 120 MM
SDP
Supplier:  Bellco Glass, Inc. 452800904
Encompass_Preferred
Borosil Watch Glass, S-Line Soda Glass, 120 mm, CS/20Borosil Watch Glasses (S-Line) are made of soda glass and each is molded to the same radius of curvature for convenient stacking. The watch glasses are high clarity and free of bubbles. A special process is used that results in an ideal flat curved surface. Watch glasses have many usages from being a surface to evaporate a liquid on, to holding solids while being weighed, for heating small amounts of substances or even as just a cover for a beaker. Watch glasses are nice to have on hand for they are versatile in a laboratory setting. Material: Soda GlassColor: ClearCapacity: 120mmClass/ Quality Grade: Type IIQuantity Per Case: 20
Catalog No. 50-254-9858