Learn More
Bellco Glass, Inc. Borosil Weighing Bottles with Interchangeable Stopper 20 mL
SDP
Supplier:  Bellco Glass, Inc. 452800709
Encompass_Preferred
Borosil Weighing Bottles with Interchangeable Stopper, 20 mL, CS/10Borosil Weighing Bottles with interchangeable stoppers are mechanically strong and chemically resistant. Weighing bottles are used for the precise weighing of solids. These bottles come with a glass interchangeable stopper which seals the inner contents to prevent spilling. Each bottle is manufactured from Class A quality grade 3.3 Borosilicate glass.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 20mLApprox O.D. x Height: 50mm x 35mmNeck Type: StopperClass/ Quality Grade: Type IQuantity Per Case: 10IS 1574
Catalog No. 50-255-0053