Learn More
Bellco Glass, Inc. Borosil Weighing Bottles with Interchangeable Stopper 5 mL
SDP
Catalog No. 502550055
Encompass_Preferred
Borosil Weighing Bottles with Interchangeable Stopper, 5 mL, CS/30Borosil Weighing Bottles with interchangeable stoppers are mechanically strong and chemically resistant. Weighing bottles are used for the precise weighing of solids. These bottles come with a glass interchangeable stopper which seals the inner contents to prevent spilling. Each bottle is manufactured from Class A quality grade 3.3 Borosilicate glass.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 5mLApprox O.D. x Height: 20mm x 40mmNeck Type: StopperClass/ Quality Grade: Type IQuantity Per Case: 30IS 1574
Catalog No. 50-255-0055 Supplier Bellco Glass, Inc. Supplier No. 452800707
Borosil Weighing Bottles with Interchangeable Stopper, 5 mL, CS/30Borosil Weighing Bottles with interchangeable stoppers are mechanically strong and chemically resistant. Weighing bottles are used for the precise weighing of solids. These bottles come with a glass interchangeable stopper which seals the inner contents to prevent spilling. Each bottle is manufactured from Class A quality grade 3.3 Borosilicate glass.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 5mLApprox O.D. x Height: 20mm x 40mmNeck Type: StopperClass/ Quality Grade: Type IQuantity Per Case: 30IS 1574