Learn More
Bellco Glass, Inc. Borosil Weighing Bottles with Interchangeable Stopper 60 mL
SDP
Catalog No. 502550050
Encompass_Preferred
Borosil Weighing Bottles with Interchangeable Stopper, 60 mL, CS/5Borosil Weighing Bottles with interchangeable stoppers are mechanically strong and chemically resistant. Weighing bottles are used for the precise weighing of solids. These bottles come with a glass interchangeable stopper which seals the inner contents to prevent spilling. Each bottle is manufactured from Class A quality grade 3.3 Borosilicate glass.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 60mLApprox O.D. x Height: 40mm x 90mmNeck Type: StopperClass/ Quality Grade: Type IQuantity Per Case: 5IS 1574
Catalog No. 50-255-0050 Supplier Bellco Glass, Inc. Supplier No. 452800712
Borosil Weighing Bottles with Interchangeable Stopper, 60 mL, CS/5Borosil Weighing Bottles with interchangeable stoppers are mechanically strong and chemically resistant. Weighing bottles are used for the precise weighing of solids. These bottles come with a glass interchangeable stopper which seals the inner contents to prevent spilling. Each bottle is manufactured from Class A quality grade 3.3 Borosilicate glass.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 60mLApprox O.D. x Height: 40mm x 90mmNeck Type: StopperClass/ Quality Grade: Type IQuantity Per Case: 5IS 1574