Learn More
Bellco Glass, Inc. Borosil Culture Tubes Media Round Bottom PP cap with Liner 10 mL
SDP
Supplier:  Bellco Glass, Inc. 452800885
Encompass_Preferred
Borosil Media Culture Tubes, Round Bottom, PP Screw Cap with PTFE Liner, 10 mL, CS/100Borosil Media Culture Tubes with a Round Bottom are the ideal replacement for media culture bottles. The polypropylene screw cap has a PTFE liner to help seal and secure the inner contents inside the culture tubes. The caps are specially designed to facilitate the handling and sealing when in use. Material: 3.3 Borosilicate GlassColor: ClearCapacity: 10mLApprox O.D. x Length: 16mm x 125mmClass/ Quality Grade: Type I, Class AQuantity Per Case: 100
Catalog No. 50-254-9877