Learn More
Thermo Scientific Emitters, borosilicate, medium (0.8 mm tip), 52 mm length, Au/Pd single coating (100 pieces)
Catalog No. NC0355451 Shop All Thermo Scientific Products
Encompass
Borosilicate glass tubes with narrow tip for accurate dispensing by capillary action of gases or liquids in laboratory equipment, withstanding temperatures up to 500°C due to high heat resistance.
Catalog No. NC0355451 Supplier Thermo Scientific Supplier No. ES380
Borosilicate glass tubes with narrow tip for accurate dispensing by capillary action of gases or liquids in laboratory equipment, withstanding temperatures up to 500°C due to high heat resistance.