Learn More
Biosurface Technologies Corp Borosilicate Glass Disc Coupon, 0.5 inch diameter for use with CDC, IBR, and RDR Biofilm Reactors
Catalog No. NC1727328
Encompass
This 0.5 inch (12.7mm) diameter coupon is manufactured from borosilicate glass, and is designed for biofilm or biofilm-related studies in a laboratory environment using the CDC Biofilm Reactor, Bio-inLine Biofilm Reactor, or the Rotating Disk Biofilm Reactor. This item has a minimum quantity of 6 per supplier requirements.
Catalog No. NC1727328 Supplier Biosurface Technologies Corp Supplier No. RD128GL
This 0.5 inch (12.7mm) diameter coupon is manufactured from borosilicate glass, and is designed for biofilm or biofilm-related studies in a laboratory environment using the CDC Biofilm Reactor, Bio-inLine Biofilm Reactor, or the Rotating Disk Biofilm Reactor. This item has a minimum quantity of 6 per supplier requirements.