Learn More
Alkali Scientific Borosilicate Glass Culture Tubes (DCTs) 1000/Case, 10x75mm (4mL)
SDP
Catalog No. 5000662642
Encompass_Preferred
Alkali Scientific™ offers high quality borosilicate glass culture tubes designed for reliable performance in clinical, research, and industrial laboratories. Manufactured from chemically resistant borosilicate glass, these tubes minimize pH shifts and contamination risk, making them suitable for sensitive applications such as tissue culture, blood banking, and sample storage. Each tube features a smooth fire polished rim and rounded bottom for safe handling and consistent experimental results. Rigid dimensional standards ensure compatibility with common racks, centrifuges, and laboratory automation systems. Tubes are packaged in four shrink wrapped boxes of 250 tubes each (1,000 total) to maintain cleanliness, protect from particles, and support efficient laboratory storage and dispensing.
Catalog No. 50-006-62642 Supplier Alkali Scientific Supplier No. BCT04
Alkali Scientific™ offers high quality borosilicate glass culture tubes designed for reliable performance in clinical, research, and industrial laboratories. Manufactured from chemically resistant borosilicate glass, these tubes minimize pH shifts and contamination risk, making them suitable for sensitive applications such as tissue culture, blood banking, and sample storage. Each tube features a smooth fire polished rim and rounded bottom for safe handling and consistent experimental results. Rigid dimensional standards ensure compatibility with common racks, centrifuges, and laboratory automation systems. Tubes are packaged in four shrink wrapped boxes of 250 tubes each (1,000 total) to maintain cleanliness, protect from particles, and support efficient laboratory storage and dispensing.