Learn More
Alkali Scientific Borosilicate Glass Culture Tubes (DCTs) 1000/Case, 12x75mm (6mL)
SDP
Catalog No. 5000662300
Encompass_Preferred
Alkali Scientific™ borosilicate glass culture tubes are designed for dependable use in clinical, research, and industrial laboratory environments. Constructed from chemically durable borosilicate glass, these tubes help minimize pH variation and reduce contamination risks during sensitive laboratory procedures including tissue culture, blood banking, and sample storage. Each tube features a fire polished rim and rounded bottom that improve safety and ease of handling while promoting consistent laboratory results. Strict dimensional tolerances ensure compatibility with common racks, centrifuges, and automated lab systems. The tubes are packaged in four shrink wrapped boxes of 250 tubes each (1,000 total) to maintain cleanliness and ensure efficient storage and dispensing in laboratory settings.
Catalog No. 50-006-62300 Supplier Alkali Scientific Supplier No. BCT06
Alkali Scientific™ borosilicate glass culture tubes are designed for dependable use in clinical, research, and industrial laboratory environments. Constructed from chemically durable borosilicate glass, these tubes help minimize pH variation and reduce contamination risks during sensitive laboratory procedures including tissue culture, blood banking, and sample storage. Each tube features a fire polished rim and rounded bottom that improve safety and ease of handling while promoting consistent laboratory results. Strict dimensional tolerances ensure compatibility with common racks, centrifuges, and automated lab systems. The tubes are packaged in four shrink wrapped boxes of 250 tubes each (1,000 total) to maintain cleanliness and ensure efficient storage and dispensing in laboratory settings.