Learn More
Alkali Scientific Borosilicate Glass Culture Tubes (DCTs) 1000/Case, 13x100mm (10mL)
SDP
Catalog No. 5000662641
Encompass_Preferred
Alkali Scientific™ borosilicate glass culture tubes provide reliable performance for clinical, research, and industrial laboratory applications. Made from high quality chemically resistant borosilicate glass, these tubes reduce pH changes and help prevent contamination, making them ideal for sensitive procedures such as tissue culture, blood banking, and sample storage. Each tube includes a smooth fire polished rim and rounded bottom to support safe handling and consistent experimental results. Precise manufacturing standards ensure compatibility with standard laboratory racks, centrifuges, and automated systems. Tubes are packaged in four shrink wrapped boxes of 250 tubes each (1,000 total) to maintain product cleanliness, reduce particle exposure, and simplify laboratory storage and dispensing.
Catalog No. 50-006-62641 Supplier Alkali Scientific Supplier No. BCT010
Alkali Scientific™ borosilicate glass culture tubes provide reliable performance for clinical, research, and industrial laboratory applications. Made from high quality chemically resistant borosilicate glass, these tubes reduce pH changes and help prevent contamination, making them ideal for sensitive procedures such as tissue culture, blood banking, and sample storage. Each tube includes a smooth fire polished rim and rounded bottom to support safe handling and consistent experimental results. Precise manufacturing standards ensure compatibility with standard laboratory racks, centrifuges, and automated systems. Tubes are packaged in four shrink wrapped boxes of 250 tubes each (1,000 total) to maintain product cleanliness, reduce particle exposure, and simplify laboratory storage and dispensing.