Learn More
Alkali Scientific Borosilicate Glass Culture Tubes (DCTs) 1000/Case, 16x100mm (14mL)
SDP
Catalog No. 5000662824
Encompass_Preferred
Alkali Scientific™ borosilicate glass culture tubes are engineered for reliable performance across clinical, research, and industrial laboratory environments. Produced from chemically resistant borosilicate glass, these tubes minimize pH fluctuations and help reduce contamination risk during sensitive laboratory processes such as tissue culture, blood banking, and sample storage. A smooth fire polished rim and rounded bottom support safe handling and consistent laboratory performance. Tight dimensional tolerances ensure compatibility with standard racks, centrifuges, and laboratory automation systems. Tubes are packaged in four shrink wrapped boxes of 250 tubes each (1,000 total) to preserve cleanliness, protect from contamination, and allow convenient storage and dispensing in laboratory workflows.
Catalog No. 50-006-62824 Supplier Alkali Scientific Supplier No. BCT014
Alkali Scientific™ borosilicate glass culture tubes are engineered for reliable performance across clinical, research, and industrial laboratory environments. Produced from chemically resistant borosilicate glass, these tubes minimize pH fluctuations and help reduce contamination risk during sensitive laboratory processes such as tissue culture, blood banking, and sample storage. A smooth fire polished rim and rounded bottom support safe handling and consistent laboratory performance. Tight dimensional tolerances ensure compatibility with standard racks, centrifuges, and laboratory automation systems. Tubes are packaged in four shrink wrapped boxes of 250 tubes each (1,000 total) to preserve cleanliness, protect from contamination, and allow convenient storage and dispensing in laboratory workflows.