Learn More
Alkali Scientific Borosilicate Glass Test Tubes, 16x150mm, Round Bottom, 23ml VOL, 25/Pk, 1000/Case
SDP
Catalog No. 503027038
Encompass_Preferred
These 16x150mm Glass Test Tubes are precision-engineered for laboratory reliability and performance. Made from high-quality borosilicate glass, they offer exceptional resistance to thermal shock, chemical corrosion, and mechanical stress, ensuring dependable results in various scientific applications. Material: Borosilicate glass for superior durability and clarity Design: Round bottom for uniform heating and ease of handling Capacity: 23ml volume, ideal for small to medium-scale experiments Packaging: Inner pack of 25 tubes, 1000 tubes per case for bulk usage These test tubes are perfect for use in clinical, industrial, and academic laboratories, offering consistent performance in chemical reactions, sample storage, and heating procedures. Whether you're conducting routine lab work or advanced research, these glass test tubes are a dependable choice.
Catalog No. 50-302-7038 Supplier Alkali Scientific Supplier No. KS212
These 16x150mm Glass Test Tubes are precision-engineered for laboratory reliability and performance. Made from high-quality borosilicate glass, they offer exceptional resistance to thermal shock, chemical corrosion, and mechanical stress, ensuring dependable results in various scientific applications. Material: Borosilicate glass for superior durability and clarity Design: Round bottom for uniform heating and ease of handling Capacity: 23ml volume, ideal for small to medium-scale experiments Packaging: Inner pack of 25 tubes, 1000 tubes per case for bulk usage These test tubes are perfect for use in clinical, industrial, and academic laboratories, offering consistent performance in chemical reactions, sample storage, and heating procedures. Whether you're conducting routine lab work or advanced research, these glass test tubes are a dependable choice.