Learn More
Sutter Instruments BOROSILICATE TB 1MM THIN WALL
SDP
Catalog No. NC9626010
Encompass_Preferred
Standard Wall Borosilicate Tubing (Without Filament), Pieces per package:225, Outside Diameter :1.00mm, Inside Diameter : 0.75mm, Overall Length: 15cm, Borosilicate Glass
Catalog No. NC9626010 Supplier Sutter Instruments Supplier No. FGGB1007515
Standard Wall Borosilicate Tubing (Without Filament), Pieces per package:225, Outside Diameter :1.00mm, Inside Diameter : 0.75mm, Overall Length: 15cm, Borosilicate Glass