Learn More
Chemglass Life Sciences Chemglass Borosilicate Tubing for Swagelok Compression Fittings - 1/4 in OD x 3.8 mm ID x 16 in
SDP
Catalog No. 502733519
Encompass_Preferred
Chemglass borosilicate tubing engineered for use with PTFE ferrule compression fittings. Precision ground outer diameter and tight tolerances provide reliable, leak-resistant connections for laboratory fluid and gas lines.
  • Material: Borosilicate glass.
  • Outer diameter: 1/4 in ±0.002 in (6.35 mm ±0.05 mm).
  • Inner diameter: 3.8 mm (0.15 in).
  • Length: 16 in (406 mm).
  • Precision ground outer diameter for consistent sealing with PTFE ferrule compression fittings.
  • Made in the United States.
Catalog No. 50-273-3519 Supplier Chemglass Life Sciences Supplier No. CG71301
Chemglass borosilicate tubing engineered for use with PTFE ferrule compression fittings. Precision ground outer diameter and tight tolerances provide reliable, leak-resistant connections for laboratory fluid and gas lines.
  • Material: Borosilicate glass.
  • Outer diameter: 1/4 in ±0.002 in (6.35 mm ±0.05 mm).
  • Inner diameter: 3.8 mm (0.15 in).
  • Length: 16 in (406 mm).
  • Precision ground outer diameter for consistent sealing with PTFE ferrule compression fittings.
  • Made in the United States.