Learn More
Research Products International Corp Borosilicate Glass Shell Vials, 5ml Capacity, Natural Polyethylene Stopper, 1152 per Case
SDP
Catalog No. 50213000
Encompass
Manufactured from Type 1 borosilicate glass. Supplied with a polyethylene stopper having double sealing rings for a leak-proof seal.
Catalog No. 50-213-000 Supplier Research Products International Corp Supplier No. 125502
Manufactured from Type 1 borosilicate glass. Supplied with a polyethylene stopper having double sealing rings for a leak-proof seal.