Learn More
Research Products International Corp Low Background Borosilicate Glass Vials, 8ml Capacity, Poly Lined Plastic Screw Caps, White, 1000 per Case
SDP
Catalog No. 50550152
Encompass
Manufactured from low background borosilicate glass selected for best performance in liquid scintillation counting. Special attention given to all critical dimensions such as wall thickness, vial diameter and length.
Catalog No. 50-550-152 Supplier Research Products International Corp Supplier No. 125505
Manufactured from low background borosilicate glass selected for best performance in liquid scintillation counting. Special attention given to all critical dimensions such as wall thickness, vial diameter and length.