Learn More
Creative Biomart Recombinant Borrelia Bavarriensis 58 Antigen, His-tagged
Supplier:  Creative Biomart BB5008
Encompass_Preferred
Recombinant Borrelia Bavarriens 58 produced in E. coli is a non-glycosylated, polypeptide chain having a calculated molecular mass of 61 kDa.Borrelia Bavarriensis 58 is expressed with a 6 His tag a
Catalog No. 50-208-4386