Learn More
Creative Biomart Recombinant Borrelia Garinii OspC Antigen, His-tagged
Supplier:  Creative Biomart BGO013
Encompass_Preferred
Recombinant Borrelia Garinii Outer Surface Protein C produced in E. coli is a non-glycosylated, polypeptide chain having a calculated molecular mass of 21,672 Dalton.Borrelia Garinii OspC is expressed
Catalog No. 50-208-4391