Learn More
Creative Biomart Recombinant Borrelia Miyamotoi GlpQ Antigen, His-tagged
Catalog No. 502084392
Encompass_Preferred
Recombinant Borrelia Miyamotoi GlpQ produced in E. coli is a non-glycosylated, polypeptide chain having a calculated molecular mass of 38,132 Dalton.Borrelia Miyamotoi GlpQ is expressed with a 10 H
Catalog No. 50-208-4392 Supplier Creative Biomart Supplier No. BMG014
Recombinant Borrelia Miyamotoi GlpQ produced in E. coli is a non-glycosylated, polypeptide chain having a calculated molecular mass of 38,132 Dalton.Borrelia Miyamotoi GlpQ is expressed with a 10 H