Learn More
Creative Biomart Recombinant Borrelia Burgdorferi (strain B31) OspA Antigen, His-tagged
Catalog No. 502084394
Encompass_Preferred
Recombinant Borrelia Burgdorferi (strain B31) OspA produced in E. coli is a non-glycosylated, full-length polypeptide chain having a calculated molecular mass of 27 kDa.Borrelia OspA is expressed with
Catalog No. 50-208-4394 Supplier Creative Biomart Supplier No. BOA016
Recombinant Borrelia Burgdorferi (strain B31) OspA produced in E. coli is a non-glycosylated, full-length polypeptide chain having a calculated molecular mass of 27 kDa.Borrelia OspA is expressed with