Learn More
Creative Biomart Recombinant Borrelia p100 Antigen, His-tagged
Catalog No. 502084396
Encompass_Preferred
Recombinant Borrelia Burgdorferi p100 (p100/p83) produced in SF9 is a glycosylated, polypeptide chain having a calculated molecular mass of 77,813 Dalton.Borrelia p100 is expressed with a 6 His tag
Catalog No. 50-208-4396 Supplier Creative Biomart Supplier No. BP0018
Recombinant Borrelia Burgdorferi p100 (p100/p83) produced in SF9 is a glycosylated, polypeptide chain having a calculated molecular mass of 77,813 Dalton.Borrelia p100 is expressed with a 6 His tag