Learn More
Creative Biomart Recombinant Borrelia p41 Antigen, His-tagged
Supplier:  Creative Biomart BPS020
Encompass_Preferred
Recombinant Borrelia Burgdorferi p41 produced in SF9 is a glycosylated, polypeptide chain having a calculated molecular mass of 36,578 Dalton.Borrelia p41 is expressed with a 6 His tag at N-terminu
Catalog No. 50-208-4398