Learn More
Creative Biomart Recombinant Borrelia p45 Antigen, His-tagged
Supplier:  Creative Biomart BP5021
Encompass_Preferred
Recombinant Borrelia Burgdorferi p45 produced in SF9 is a glycosylated, polypeptide chain having a calculated molecular mass of 45,259.3 Dalton.Borrelia p45 is expressed with a 10 His tag at N-term
Catalog No. 50-208-4399