Learn More
Creative Biomart Recombinant Borrelia p66 Antigen, His-tagged
Supplier:  Creative Biomart BP6023
Encompass_Preferred
Recombinant Borrelia Burgdorferi p66 produced in E. coli is a non-glycosylated, polypeptide chain having a calculated molecular mass of 66 kDa. Borrelia p66 is expressed with a 10 His tag at N-term
Catalog No. 50-208-4401