Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borrelidin, Tocris Bioscience™

Catalog No. 47061
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-061 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 47-061 Supplier R&D Systems Supplier No. 4706/1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Antiangiogenic; inhibits threonyl-tRNA synthetase

Antibiotic and antiangiogenic. Selectively inhibits threonyl-tRNA synthetase (ThrRS). Exhibits antiangiogenic activity in a mouse model of tumor angiogenesis. Induces apoptosis in capillary tube-forming endothelial cells; disrupts capillary tubes and inhibits their formation (IC50 = 0.8 nM in rat aorta). Induces the unfolded protein response (UPR) and apoptosis in oral cancer cells.

Chemical Identifiers

CAS 7184-60-3
Molecular Formula C28H43NO6
Molecular Weight (g/mol) 489.653
InChI Key OJCKRNPLOZHAOU-MJQQLCDASA-N
Synonym treponemycin
PubChem CID 133655849
IUPAC Name (1S,2S)-2-[(2R,4Z,6E,8S,9R,13R,15R,16R)-7-cyano-8,16-dihydroxy-9,11,13,15-tetramethyl-18-oxo-1-oxacyclooctadeca-4,6-dien-2-yl]cyclopentane-1-carboxylic acid
SMILES CC1CC(CC(C(C(=CC=CCC(OC(=O)CC(C(C1)C)O)C2CCCC2C(=O)O)C#N)O)C)C

Specifications

Quantity 1 mg
Formula Weight 489.64
Percent Purity >98%
Chemical Name or Material Borrelidin
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.