Learn More
Medchemexpress LLC HY-10227 100mg , Bortezomib CAS:179324-69-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10227100MG
Encompass_Preferred
HY-10227 100mg Bortezomib CAS: 179324-69-7 Bortezomib (PS-341) is a reversible and selective proteasome inhibitor, and potently inhibits 20S proteasome (Ki=0.6 nM) by targeting a threonine residue. Bortezomib disrupts the cell cycle, induces apoptosis, and inhibits NF-κB. Bortezomib is the first proteasome inhibitor anticancer agent. Anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9295