Learn More
TARGETMOL CHEMICALS INC BORTEZOMIB 100MG
SDP
Catalog No. 502580836
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bortezomib (Brotezamide) (PS-341) is a potent 20S proteasome inhibitor (Ki 0.6 nM in a cell-free assay). purity: 99%
Catalog No. 50-258-0836 Supplier TARGETMOL CHEMICALS INC Supplier No. T2399
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bortezomib (Brotezamide) (PS-341) is a potent 20S proteasome inhibitor (Ki 0.6 nM in a cell-free assay). purity: 99%