Learn More
Abcam Bortezomib, proteasome inhibitor, 5MG
Catalog No. NC1539114
Encompass_Preferred
MW 384.2 Da, Purity >98%. Potent, selective, reversible proteasome inhibitor (Ki = 0.6 nM). Inhibits proliferation of a number of tumor cell lines (IC₅₀ = 7 nM). Inhibits TNF synthesis and FGF-induced angiogenesis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1539114 Supplier Abcam Supplier No. AB1421235MG
MW 384.2 Da, Purity >98%. Potent, selective, reversible proteasome inhibitor (Ki = 0.6 nM). Inhibits proliferation of a number of tumor cell lines (IC₅₀ = 7 nM). Inhibits TNF synthesis and FGF-induced angiogenesis.
The product is subject to the following: Abcam Restricted Use Statement