Learn More
Medchemexpress LLC Bortezomib-d8 | 00-00-0 | MFCD09056737 | 98.0% | 392.29 g/mol | C19H17D8BN4O4 | 1 MG
SDP
Catalog No. 5000245892
Encompass_Preferred
Bortezomib-d8 is the deuterium-labeled (d8) isotopologue of bortezomib, a reversible and selective 20S proteasome inhibitor used in research on proteasome function, apoptosis, and multiple myeloma. It is supplied as a high-purity analytical reagent for use in mass spectrometry, tracer studies, and mechanistic experiments.
  • Deuterium-labeled analogue for use as an internal standard.
  • High purity suitable for analytical applications.
  • Optimized for mass spectrometry and quantitative assays.
  • Small package size for low-quantity, high-value experiments.
  • Stable isotope labeling aids in accurate tracer and mechanistic studies.
Catalog No. 50-002-45892 Supplier Medchemexpress LLC Supplier No. HY10227S1MG
Bortezomib-d8 is the deuterium-labeled (d8) isotopologue of bortezomib, a reversible and selective 20S proteasome inhibitor used in research on proteasome function, apoptosis, and multiple myeloma. It is supplied as a high-purity analytical reagent for use in mass spectrometry, tracer studies, and mechanistic experiments.
  • Deuterium-labeled analogue for use as an internal standard.
  • High purity suitable for analytical applications.
  • Optimized for mass spectrometry and quantitative assays.
  • Small package size for low-quantity, high-value experiments.
  • Stable isotope labeling aids in accurate tracer and mechanistic studies.