Learn More
Cayman Chemical BortezomIb ImpurIty A 100mg
SDP
Catalog No. 502912097
Encompass_Preferred
A metabolite of bortezomib; a degradation product of bortezomib formed under oxidative conditions
Catalog No. 50-291-2097 Supplier Cayman Chemical Supplier No. 36499100 MG
A metabolite of bortezomib; a degradation product of bortezomib formed under oxidative conditions