Learn More
Cayman Chemical BortezomIb ImpurIty A 10mg
SDP
Catalog No. 502912903
Encompass_Preferred
A metabolite of bortezomib; a degradation product of bortezomib formed under oxidative conditions
Catalog No. 50-291-2903 Supplier Cayman Chemical Supplier No. 3649910 MG
A metabolite of bortezomib; a degradation product of bortezomib formed under oxidative conditions