Learn More
Cayman Chemical BortezomIb ImpurIty A 50mg
SDP
Catalog No. 502912127
Encompass_Preferred
A metabolite of bortezomib; a degradation product of bortezomib formed under oxidative conditions
Catalog No. 50-291-2127 Supplier Cayman Chemical Supplier No. 3649950 MG
A metabolite of bortezomib; a degradation product of bortezomib formed under oxidative conditions