Learn More
Apexbio Technology LLC Bortezomib (PS-341) 179324-69-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063252
Encompass_Preferred
Bortezomib (PS-341 CAS number 179324-69-7) is a reversible proteasome inhibitor structurally composed as an N-terminally protected dipeptide (Pyz-Phe-boroLeu) containing pyrazinoic acid phenylalanine and leucine with boronic acid substitution It exerts biological activity primarily by inhibiting proteasomal degradation pathways thereby accumulating pro-apoptotic factors and initiating programmed cell death Bortezomib inhibits proliferation in cell-based assays such as human non-small cell lung cancer H460 cells (IC50 0 1 M) It has clinical approval for relapsed multiple myeloma and mantle cell lymphoma and is widely employed in research to study proteasome-regulated cellular processes and apoptosis signaling pathways
Catalog No. 50-000-63252 Supplier Apexbio Technology LLC Supplier No. A26145.1
Bortezomib (PS-341 CAS number 179324-69-7) is a reversible proteasome inhibitor structurally composed as an N-terminally protected dipeptide (Pyz-Phe-boroLeu) containing pyrazinoic acid phenylalanine and leucine with boronic acid substitution It exerts biological activity primarily by inhibiting proteasomal degradation pathways thereby accumulating pro-apoptotic factors and initiating programmed cell death Bortezomib inhibits proliferation in cell-based assays such as human non-small cell lung cancer H460 cells (IC50 0 1 M) It has clinical approval for relapsed multiple myeloma and mantle cell lymphoma and is widely employed in research to study proteasome-regulated cellular processes and apoptosis signaling pathways