Learn More
Selleck Chemical LLC Bortezomib (PS-341) 25mg 179324-69-7 LDP-341, MLM341
SDP
Catalog No. 507416
Encompass_Preferred
Bortezomib (PS-341, LDP-341, MLM341) is a potent 20S proteasome inhibitor with Ki of 0.6 nM. It exhibits favorable selectivity towards tumor cells over normal cells. Bortezomib (PS-341) inhibits NF-?B and induces ERK phosphorylation to suppress cathepsin B and inhibit the catalytic process of autophagy in ovarian cancer and other solid tumors. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-741-6 Supplier Selleck Chemical LLC Supplier No. S101325MG
Bortezomib (PS-341, LDP-341, MLM341) is a potent 20S proteasome inhibitor with Ki of 0.6 nM. It exhibits favorable selectivity towards tumor cells over normal cells. Bortezomib (PS-341) inhibits NF-?B and induces ERK phosphorylation to suppress cathepsin B and inhibit the catalytic process of autophagy in ovarian cancer and other solid tumors. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.