Learn More
Medchemexpress LLC Borussertib | 1800070-77-2 | 99.2% | 1 MG

Supplier: Medchemexpress LLC HY1229131MG
Borussertib is a covalent-allosteric, first-in-class inhibitor of protein kinase Akt used as a chemical probe in research. It has an IC50 of 0.8 nM and a Ki of 2.2 nM for Akt, molecular weight 596.68, and chemical formula C36H32N6O3. It is supplied in small research pack sizes and is soluble in DMSO.
- Covalent-allosteric Akt inhibitor with IC50 0.8 nM and Ki 2.2 nM.
- Chemical probe suited for in vitro and in vivo studies.
- High supplied purity (99.18%).
- Soluble in DMSO (25 mg/mL) and formulatable for in vivo use.
- Stable when stored as powder at -20°C (long-term) or 4°C (short-term).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.