Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Borussertib | 1800070-77-2 | 99.2% | 25 MG
SDP

Catalog No. 5000230585
Encompass_Preferred

Borussertib is a covalent-allosteric, first-in-class small-molecule inhibitor of protein kinase Akt used as a chemical probe in preclinical research. It demonstrates sub-nanomolar potency against wild-type Akt and is supplied as a high-purity solid suitable for in vitro and in vivo studies.

  • Sub-nanomolar Akt inhibitor (IC50 0.8 nM; Ki 2.2 nM).
  • Designed as a covalent-allosteric chemical probe for Akt signaling studies.
  • High reported purity: 99.18%.
  • Molecular formula C36H32N6O3; molecular weight 596.68 g/mol.
  • Soluble in DMSO (25 mg/mL); recommended in vivo formulations provided.
  • Powder storage: -20 °C for long-term stability; follow solvent storage recommendations.

Catalog No. 50-002-30585 Supplier Medchemexpress LLC Supplier No. HY12291325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Borussertib is a covalent-allosteric, first-in-class small-molecule inhibitor of protein kinase Akt used as a chemical probe in preclinical research. It demonstrates sub-nanomolar potency against wild-type Akt and is supplied as a high-purity solid suitable for in vitro and in vivo studies.

  • Sub-nanomolar Akt inhibitor (IC50 0.8 nM; Ki 2.2 nM).
  • Designed as a covalent-allosteric chemical probe for Akt signaling studies.
  • High reported purity: 99.18%.
  • Molecular formula C36H32N6O3; molecular weight 596.68 g/mol.
  • Soluble in DMSO (25 mg/mL); recommended in vivo formulations provided.
  • Powder storage: -20 °C for long-term stability; follow solvent storage recommendations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.