Learn More
Selleck Chemical LLC Borussertib-5mg
SDP
Supplier:  Selleck Chemical LLC S88395MG
Encompass_Preferred
Borussertib is a covalent-allosteric inhibitor of protein kinase Akt with an IC50 of 0.8 nM and a Ki of 2.2 nM for WT Akt.
Catalog No. 50-238-9861