Learn More
Medchemexpress LLC Boscalid | 188425-85-6 | 100 MG
SDP
Catalog No. 5000232415
Encompass_Preferred
Boscalid (CAS 188425-85-6) is a pyridine carboxamide fungicide supplied as an analytical reference standard for use in analytical and research applications. It acts as a succinate dehydrogenase (SDH) inhibitor and is used for method development, identity confirmation, and quantitative analysis in chromatographic and mass spectrometric workflows.
  • Used as an analytical/reference standard for HPLC, GC, and MS.
  • Suitable for identity confirmation and quantitative calibration in analytical workflows.
  • Applicable for residue analysis and method validation.
  • Molecular formula: C18H12Cl2N2O; molecular weight: 343.21.
  • Supplied in small pack sizes appropriate for laboratory analytical use.
Catalog No. 50-002-32415 Supplier Medchemexpress LLC Supplier No. HY119976100MG
Boscalid (CAS 188425-85-6) is a pyridine carboxamide fungicide supplied as an analytical reference standard for use in analytical and research applications. It acts as a succinate dehydrogenase (SDH) inhibitor and is used for method development, identity confirmation, and quantitative analysis in chromatographic and mass spectrometric workflows.
  • Used as an analytical/reference standard for HPLC, GC, and MS.
  • Suitable for identity confirmation and quantitative calibration in analytical workflows.
  • Applicable for residue analysis and method validation.
  • Molecular formula: C18H12Cl2N2O; molecular weight: 343.21.
  • Supplied in small pack sizes appropriate for laboratory analytical use.