Learn More
Medchemexpress LLC Boscalid | 188425-85-6 | 99.8% | 1 G
SDP
Catalog No. 5000232284
Encompass_Preferred
Boscalid is a fungicidal research chemical that inhibits succinate dehydrogenase (SDH), disrupting fungal respiration. It is supplied for research and analytical applications and is available as a solid or as a 10 mM solution in DMSO.
  • Provides fungicidal activity as a succinate dehydrogenase inhibitor
  • High purity suitable for analytical use (99.8%)
  • Molecular formula C18H12Cl2N2O; molecular weight 343.21 g/mol
  • Available in solid form and as a 10 mM solution in DMSO; common pack sizes include 50 mg-1 g
  • Intended for research use only; not for human or veterinary use
Catalog No. 50-002-32284 Supplier Medchemexpress LLC Supplier No. HY1199761G
Boscalid is a fungicidal research chemical that inhibits succinate dehydrogenase (SDH), disrupting fungal respiration. It is supplied for research and analytical applications and is available as a solid or as a 10 mM solution in DMSO.
  • Provides fungicidal activity as a succinate dehydrogenase inhibitor
  • High purity suitable for analytical use (99.8%)
  • Molecular formula C18H12Cl2N2O; molecular weight 343.21 g/mol
  • Available in solid form and as a 10 mM solution in DMSO; common pack sizes include 50 mg-1 g
  • Intended for research use only; not for human or veterinary use