Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boscalid | 188425-85-6 | 99.8% | 50 MG
SDP

Catalog No. 5000234069
Encompass_Preferred

Boscalid is a succinate dehydrogenase (SDH) inhibitor fungicide supplied as an analytical standard for research use. It is provided as a powder suitable for preparing stock solutions and biochemical assays, intended for research applications only.

  • Analytical standard for research applications.
  • Succinate dehydrogenase (SDH) inhibitor fungicide.
  • High purity (99.8%) for reliable analytical results.
  • Molecular weight 343.21 g/mol; formula C18H12Cl2N2O.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-34069 Supplier Medchemexpress LLC Supplier No. HY11997650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boscalid is a succinate dehydrogenase (SDH) inhibitor fungicide supplied as an analytical standard for research use. It is provided as a powder suitable for preparing stock solutions and biochemical assays, intended for research applications only.

  • Analytical standard for research applications.
  • Succinate dehydrogenase (SDH) inhibitor fungicide.
  • High purity (99.8%) for reliable analytical results.
  • Molecular weight 343.21 g/mol; formula C18H12Cl2N2O.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.