Learn More
TARGETMOL CHEMICALS INC BOSENTAN 100MG
SDP
Catalog No. 502576550
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bosentan (Benzenesulfonamide) is a sulfonamide-derived competitive and specific endothelin receptor antagonist with a slightly higher affinity for the endothelin A receptor than endothelin B receptor. Bosentan blocks the action of endothelin 1 an extremely potent endogenous vasoconstrictor and bronchoconstrictor by binding to endothelin A and endothelin B receptors in the endothelium and vascular smooth muscle. Bosentan decreases both pulmonary and systemic vascular resistance and is particularly used in the treatment of pulmonary arterial hypertension. purity: 99%
Catalog No. 50-257-6550 Supplier TARGETMOL CHEMICALS INC Supplier No. T6264100MG
Also available in 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bosentan (Benzenesulfonamide) is a sulfonamide-derived competitive and specific endothelin receptor antagonist with a slightly higher affinity for the endothelin A receptor than endothelin B receptor. Bosentan blocks the action of endothelin 1 an extremely potent endogenous vasoconstrictor and bronchoconstrictor by binding to endothelin A and endothelin B receptors in the endothelium and vascular smooth muscle. Bosentan decreases both pulmonary and systemic vascular resistance and is particularly used in the treatment of pulmonary arterial hypertension. purity: 99%