Learn More
Apexbio Technology LLC Bosentan 147536-97-8 50mg
SDP
Catalog No. 5000094509
Encompass_Preferred
Bosentan (CAS 147536-97-8) is a small molecule that acts as a competitive antagonist at both endothelin-A (ET-A) and endothelin-B (ET-B) receptors By blocking the binding of endothelin-1 to these receptors bosentan modulates endothelin-mediated vasoconstriction and cellular proliferation pathways This compound is utilized in biomedical research to investigate the endothelin signaling axis and its role in cardiovascular physiology pulmonary hypertension and related pathologies Bosentan enables the study of receptor-specific effects of endothelin-1 inhibition and offers a tool for evaluating therapeutic strategies targeting the endothelin system
Catalog No. 50-000-94509 Supplier Apexbio Technology LLC Supplier No. B168250
Bosentan (CAS 147536-97-8) is a small molecule that acts as a competitive antagonist at both endothelin-A (ET-A) and endothelin-B (ET-B) receptors By blocking the binding of endothelin-1 to these receptors bosentan modulates endothelin-mediated vasoconstriction and cellular proliferation pathways This compound is utilized in biomedical research to investigate the endothelin signaling axis and its role in cardiovascular physiology pulmonary hypertension and related pathologies Bosentan enables the study of receptor-specific effects of endothelin-1 inhibition and offers a tool for evaluating therapeutic strategies targeting the endothelin system