Learn More
Medchemexpress LLC Bosentan | 147536-97-8 | 99.9% | C27H29N5O6S | 1 ML
SDP
Catalog No. 5000432323
Encompass_Preferred
Bosentan is a competitive and dual antagonist of endothelin-1 (ET) for the ETA and ETB receptors, with Ki values of 4.7 nM and 95 nM in human SMC, respectively. It is for research use only.
  • Competitive and dual antagonist of endothelin-1 (ET)
  • Acts on ETA and ETB receptors
  • Research use only
Catalog No. 50-004-32323 Supplier Medchemexpress LLC Supplier No. HYA001310MM/1ML
Bosentan is a competitive and dual antagonist of endothelin-1 (ET) for the ETA and ETB receptors, with Ki values of 4.7 nM and 95 nM in human SMC, respectively. It is for research use only.
  • Competitive and dual antagonist of endothelin-1 (ET)
  • Acts on ETA and ETB receptors
  • Research use only