Learn More
Medchemexpress LLC Bosentan | 147536-97-8 | 99.9% | C27H29N5O6S | 100 MG
SDP
Catalog No. 5000433135
Encompass_Preferred
Bosentan is a competitive and dual antagonist of endothelin-1 (ET) for the ETA and ETB receptors. It features a high purity of 99.9% and is used in various research applications. It is supplied as a solid.
  • Competitive and dual antagonist of endothelin-1 (ET)
  • Targets ETA and ETB receptors
  • Ki values of 4.7 nM and 95 nM in human SMC for ETA and ETB receptors
  • Soluble in DMSO
Catalog No. 50-004-33135 Supplier Medchemexpress LLC Supplier No. HYA0013100MG
Bosentan is a competitive and dual antagonist of endothelin-1 (ET) for the ETA and ETB receptors. It features a high purity of 99.9% and is used in various research applications. It is supplied as a solid.
  • Competitive and dual antagonist of endothelin-1 (ET)
  • Targets ETA and ETB receptors
  • Ki values of 4.7 nM and 95 nM in human SMC for ETA and ETB receptors
  • Soluble in DMSO